Assay Details
| Gene Symbol: RUFY2-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RUFY2-RET_R9R12.0 | - | 52 | 111 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RUFY2-RET fusion| Gene Symbol: RUFY2-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_RUFY2-RET_R9R12.0 | - | 52 | 111 | ||
Set Membership: |
|