Assay Details
| Gene Symbol: SMEK2-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SMEK2-ALK_S9A2.0 | - | 53 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMEK2-ALK fusion| Gene Symbol: SMEK2-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SMEK2-ALK_S9A2.0 | - | 53 | 119 | ||
Set Membership: |
|