Assay Details
| Gene Symbol: SQSTM1-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SQSTM1-NTRK2_S5N17.0 | - | 55 | 93 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SQSTM1-NTRK2 fusion| Gene Symbol: SQSTM1-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SQSTM1-NTRK2_S5N17.0 | - | 55 | 93 | ||