Assay Details
| Gene Symbol: TFG-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TFG-NTRK1_T6N10.0 | - | 69 | 81 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TFG-NTRK1 fusion| Gene Symbol: TFG-NTRK1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TFG-NTRK1_T6N10.0 | - | 69 | 81 | ||