Assay Details
| Gene Symbol: TRAF1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TRAF1-ALK_T6A20.0 | - | 70 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAF1-ALK fusion| Gene Symbol: TRAF1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TRAF1-ALK_T6A20.0 | - | 70 | 82 | ||