Assay Details
| Gene Symbol: VCL-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_VCL-NTRK2_V16N12.0 | - | 127 | 102 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VCL-NTRK2 fusion| Gene Symbol: VCL-NTRK2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_VCL-NTRK2_V16N12.0 | - | 127 | 102 | ||
Set Membership: |
|