Assay Details
| Gene Symbol: GBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134486.2 | NP_001127958.1 | 2 - 3 | 616 | 66 | |
| NM_052942.3 | NP_443174.1 | 3 - 4 | 724 | 66 | ||
| GenBank mRNA | AF288815.1 | - | 1 - 2 | 187 | 66 | |
| AF328727.1 | - | 3 - 4 | 687 | 66 | ||
| AF430642.1 | - | 3 - 4 | 725 | 66 | ||
| AF430643.1 | - | 2 - 3 | 605 | 66 | ||
| AK090479.1 | - | 1 - 2 | 3436 | 66 | ||
| AK315064.1 | - | 3 - 4 | 335 | 66 | ||
| AY358953.1 | - | 2 - 3 | 550 | 66 | ||
| BC031639.1 | - | 3 - 4 | 711 | 5173241 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002850.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inflammatory response
positive regulation of innate immune response
interleukin-1 beta secretion
protein homotetramerization
cellular response to interferon-gamma
interleukin-18 secretion
positive regulation of cytokine production involved in inflammatory response
positive regulation of NLRP3 inflammasome complex assembly
GTPase activity
protein binding
GTP binding
identical protein binding
inflammatory response
positive regulation of innate immune response
interleukin-1 beta secretion
protein homotetramerization
cellular response to interferon-gamma
interleukin-18 secretion
positive regulation of cytokine production involved in inflammatory response
positive regulation of NLRP3 inflammasome complex assembly
GTPase activity
protein binding
GTP binding
identical protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044470.hg.1 | TC01002850.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044470.hg.1 | ENST00000343435.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044470.hg.1 | ENST00000370459.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044470.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044470.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008972.hg.1 | ENST00000437128.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008972.hg.1 | ENST00000437128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210725.hg.1 | TC0100014857.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008972.hg.1 | ENST00000437128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008972.hg.1 | ENST00000437128.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000370459.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000443807.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000471171.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000481145.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | ENST00000490568.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_001134486.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014857.hg.1 | NM_052942.3 | Refseq |

GBP5
Genomic Map