Assay Details
| Gene Symbol: UGT2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252275.2 | NP_001239204.2 | 3 - 4 | 967 | 73 | |
| GenBank mRNA | AK314209.1 | - | 3 - 4 | 965 | 73 | |
More Information
Set Membership: |
|

UGT2A1
Genomic Map