Assay Details
| Gene Symbol: IL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314046.1 | NP_001300975.1 | 4 - 5 | 426 | 106 | |
| NM_001314047.1 | NP_001300976.1 | 4 - 5 | 413 | 106 | ||
| XM_017015285.1 | XP_016870774.1 | 6 - 7 | 1172 | 106 | ||
| GenBank mRNA | DA719342.1 | - | 4 - 5 | 411 | 106 | |
| DB199599.1 | - | 4 - 5 | 435 | 106 | ||
| DB266668.1 | - | 4 - 5 | 405 | 106 | ||
More Information
Set Membership: |
|

IL33
Genomic Map