Assay Details
| Gene Symbol: IL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199640.1 | NP_001186569.1 | 6 - 7 | 555 | 98 | |
| NM_001199641.1 | NP_001186570.1 | 4 - 5 | 303 | 98 | ||
| NM_001314044.1 | NP_001300973.1 | 7 - 8 | 668 | 98 | ||
| NM_001314045.1 | NP_001300974.1 | 7 - 8 | 676 | 98 | ||
| NM_001314046.1 | NP_001300975.1 | 7 - 8 | 663 | 98 | ||
| NM_001314047.1 | NP_001300976.1 | 7 - 8 | 650 | 98 | ||
| NM_001314048.1 | NP_001300977.1 | 6 - 7 | 555 | 98 | ||
| NM_033439.3 | NP_254274.1 | 7 - 8 | 681 | 98 | ||
| XM_011518061.2 | XP_011516363.1 | 6 - 7 | 616 | 98 | ||
| XM_017015285.1 | XP_016870774.1 | 9 - 10 | 1409 | 98 | ||
| GenBank mRNA | AB024518.1 | - | 6 - 7 | 620 | 98 | |
| AK295908.1 | - | 4 - 5 | 284 | 98 | ||
| AK303943.1 | - | 6 - 7 | 532 | 98 | ||
| AK313414.1 | - | 7 - 8 | 660 | 98 | ||
| AY905581.1 | - | 6 - 7 | 603 | 98 | ||
| BC047085.1 | - | 7 - 8 | 658 | 5314040 | 98 | |
| BQ878541.1 | - | 6 - 7 | 642 | 98 | ||
| CR407619.1 | - | 6 - 7 | 603 | 98 | ||
| HQ641439.1 | - | 5 - 6 | 492 | 98 | ||
More Information
Set Membership: |
|

IL33
Genomic Map