Assay Details
| Gene Symbol: SLC2A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286234.1 | NP_001273163.1 | 2 - 3 | 309 | 69 | |
| NM_001286235.1 | NP_001273164.1 | 4 - 5 | 607 | 69 | ||
| NM_001286236.1 | NP_001273165.1 | 2 - 3 | 309 | 69 | ||
| XM_005253315.3 | XP_005253372.1 | 2 - 3 | 143 | 69 | ||
| XM_005253317.4 | XP_005253374.1 | 2 - 3 | 662 | 69 | ||
| XM_011520562.1 | XP_011518864.1 | 5 - 6 | 497 | 69 | ||
| XM_011520563.2 | XP_011518865.1 | 2 - 3 | 157 | 69 | ||
| XM_011520564.2 | XP_011518866.1 | 2 - 3 | 127 | 69 | ||
| XM_017018846.1 | XP_016874335.1 | 3 - 4 | 318 | 69 | ||
| XM_017018847.1 | XP_016874336.1 | 2 - 3 | 275 | 69 | ||
| GenBank mRNA | AF481878.1 | - | 2 - 3 | 128 | 69 | |
| AK122783.1 | - | 4 - 5 | 395 | 69 | ||
| AK302881.1 | - | 2 - 3 | 122 | 69 | ||
| AY357941.1 | - | 2 - 3 | 294 | 69 | ||
| BC060766.1 | - | 2 - 3 | 156 | 5297510 | 69 | |
| BG717327.1 | - | 3 - 4 | 371 | 69 | ||
| DB061127.1 | - | 4 - 5 | 392 | 69 | ||
| DB082816.1 | - | 2 - 3 | 186 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF481878.1, AK122783.1, AK302881.1, AY357941.1, BC060766.1, NM_001286234.1, NM_001286235.1, NM_001286236.1, XM_005253315.3, XM_005253317.4, XM_011520562.1, XM_011520563.2, XM_011520564.2, XM_017018846.1, XM_017018847.1 transcripts.
Set Membership: |
|

SLC2A14
Genomic Map