Assay Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287819.1 | NP_001274748.1 | 10 - 11 | 1105 | 111 | |
| NM_001287820.1 | NP_001274749.1 | 10 - 11 | 1105 | 111 | ||
| NM_198503.3 | NP_940905.2 | 10 - 11 | 1105 | 111 | ||
| XM_006711294.2 | XP_006711357.1 | 10 - 11 | 1579 | 111 | ||
| XM_006711295.2 | XP_006711358.1 | 10 - 11 | 1579 | 111 | ||
| XM_011509482.2 | XP_011507784.1 | 9 - 10 | 1504 | 111 | ||
| XM_011509483.2 | XP_011507785.1 | 10 - 11 | 1579 | 111 | ||
| XM_017001178.1 | XP_016856667.1 | 10 - 11 | 1579 | 111 | ||
| XM_017001179.1 | XP_016856668.1 | 10 - 11 | 1579 | 111 | ||
| XM_017001180.1 | XP_016856669.1 | 10 - 11 | 1579 | 111 | ||
| XM_017001182.1 | XP_016856671.1 | 10 - 11 | 1579 | 111 | ||
| XM_017001184.1 | XP_016856673.1 | 10 - 11 | 1579 | 111 | ||
| XM_017001185.1 | XP_016856674.1 | 9 - 10 | 1504 | 111 | ||
| GenBank mRNA | AK127807.1 | - | 3 - 4 | 455 | 111 | |
| AK131498.1 | - | 9 - 10 | 1159 | 111 | ||
| AK309729.1 | - | 10 - 11 | 1094 | 111 | ||
| AY359444.1 | - | 10 - 11 | 1043 | 111 | ||
| BC103948.1 | - | 10 - 11 | 1033 | 40009146 | 111 | |
| BC103949.1 | - | 10 - 11 | 1033 | 40009147 | 111 | |
| BC103950.1 | - | 10 - 11 | 1033 | 40009148 | 111 | |
| BX647852.1 | - | 10 - 11 | 1087 | 112 | ||
| EU251076.1 | - | 10 - 11 | 983 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017001182.1, XM_017001184.1 transcripts.
Set Membership: |
|

KCNT2
Genomic Map