Assay Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287819.1 | NP_001274748.1 | 18 - 19 | 2226 | 101 | |
| NM_001287820.1 | NP_001274749.1 | 17 - 18 | 2076 | 101 | ||
| NM_198503.3 | NP_940905.2 | 18 - 19 | 2226 | 101 | ||
| XM_006711294.2 | XP_006711357.1 | 18 - 19 | 2700 | 101 | ||
| XM_006711295.2 | XP_006711358.1 | 18 - 19 | 2700 | 101 | ||
| XM_011509482.2 | XP_011507784.1 | 17 - 18 | 2625 | 101 | ||
| XM_011509483.2 | XP_011507785.1 | 18 - 19 | 2700 | 101 | ||
| XM_017001178.1 | XP_016856667.1 | 18 - 19 | 2700 | 101 | ||
| XM_017001179.1 | XP_016856668.1 | 18 - 19 | 2700 | 101 | ||
| XM_017001180.1 | XP_016856669.1 | 18 - 19 | 2700 | 101 | ||
| XM_017001181.1 | XP_016856670.1 | 8 - 9 | 1215 | 101 | ||
| XM_017001182.1 | XP_016856671.1 | 17 - 18 | 2591 | 101 | ||
| XM_017001183.1 | XP_016856672.1 | 5 - 6 | 751 | 101 | ||
| XM_017001184.1 | XP_016856673.1 | 17 - 18 | 2591 | 101 | ||
| XM_017001185.1 | XP_016856674.1 | 17 - 18 | 2625 | 101 | ||
| GenBank mRNA | AK131498.1 | - | 17 - 18 | 2301 | 101 | |
| AK309729.1 | - | 18 - 19 | 2215 | 101 | ||
| AY359444.1 | - | 18 - 19 | 2164 | 101 | ||
| BC103947.1 | - | 14 - 15 | 1607 | 101 | ||
| BC103948.1 | - | 18 - 19 | 2154 | 40009146 | 101 | |
| BC103949.1 | - | 17 - 18 | 2004 | 40009147 | 101 | |
| BC103950.1 | - | 18 - 19 | 2154 | 40009148 | 101 | |
| BX647852.1 | - | 18 - 19 | 2208 | 101 | ||
| EU251076.1 | - | 18 - 19 | 2104 | 101 | ||
More Information
Set Membership: |
|

KCNT2
Genomic Map