Assay Details
| Gene Symbol: KCNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287819.1 | NP_001274748.1 | 19 - 20 | 2399 | 116 | |
| NM_001287820.1 | NP_001274749.1 | 18 - 19 | 2249 | 116 | ||
| XM_006711295.2 | XP_006711358.1 | 19 - 20 | 2873 | 116 | ||
| XM_017001178.1 | XP_016856667.1 | 19 - 20 | 2873 | 116 | ||
| XM_017001185.1 | XP_016856674.1 | 18 - 19 | 2798 | 116 | ||
| GenBank mRNA | AK131498.1 | - | 18 - 19 | 2474 | 116 | |
| AK309729.1 | - | 19 - 20 | 2388 | 116 | ||
| BC103948.1 | - | 19 - 20 | 2327 | 40009146 | 116 | |
| BC103949.1 | - | 18 - 19 | 2177 | 40009147 | 116 | |
| BC103950.1 | - | 19 - 20 | 2327 | 40009148 | 116 | |
| BX647852.1 | - | 19 - 20 | 2381 | 116 | ||
More Information
Set Membership: |
|

KCNT2
Genomic Map