Assay Details
| Gene Symbol: FAM160A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109977.1 | NP_001103447.1 | 11 - 12 | 3132 | 69 | |
| XM_005263199.1 | XP_005263256.1 | 10 - 11 | 3007 | 69 | ||
| XM_011532220.1 | XP_011530522.1 | 9 - 10 | 2885 | 69 | ||
| XM_011532221.2 | XP_011530523.1 | 11 - 12 | 3205 | 69 | ||
| XM_011532222.2 | XP_011530524.1 | 10 - 11 | 3127 | 69 | ||
| XM_011532223.2 | XP_011530525.1 | 9 - 10 | 3002 | 69 | ||
| XM_011532224.2 | XP_011530526.1 | 9 - 10 | 2701 | 69 | ||
| XM_017008583.1 | XP_016864072.1 | 3 - 4 | 1462 | 69 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000435205.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000503146.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000505231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000508198.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000511501.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000512597.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513086.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | ENST00000513962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009001.hg.1 | NM_001109977.2 | Refseq |

FAM160A1
Genomic Map