Assay Details
| Gene Symbol: ST6GALNAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160011.1 | NP_001153483.1 | 2 - 3 | 379 | 91 | |
| NM_152996.2 | NP_694541.2 | 2 - 3 | 379 | 91 | ||
| XM_006710553.3 | XP_006710616.1 | 3 - 4 | 375 | 91 | ||
| XM_017000937.1 | XP_016856426.1 | 2 - 3 | 3521 | 91 | ||
| XM_017000938.1 | XP_016856427.1 | 2 - 3 | 3533 | 91 | ||
| XM_017000939.1 | XP_016856428.1 | 2 - 3 | 3531 | 91 | ||
| XM_017000940.1 | XP_016856429.1 | 3 - 4 | 638 | 91 | ||
| XM_017000941.1 | XP_016856430.1 | 2 - 3 | 1293 | 91 | ||
| XM_017000942.1 | XP_016856431.1 | 2 - 3 | 1098 | 91 | ||
| GenBank mRNA | AJ507291.1 | - | 2 - 3 | 237 | 91 | |
| AK091215.1 | - | 2 - 3 | 364 | 91 | ||
| AY358540.1 | - | 2 - 3 | 338 | 91 | ||
| BC039520.1 | - | 1 - 2 | 90 | 91 | ||
| BC059363.1 | - | 2 - 3 | 350 | 30341233 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017000940.1, XM_017000941.1, XM_017000942.1 transcripts.
Set Membership: |
|

ST6GALNAC3
Genomic Map