Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007769.hg.1 |
ENST00000377924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007769.hg.1 |
ENST00000377924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007769.hg.1 |
ENST00000377924.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007769.hg.1 |
NM_001195545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007769.hg.1 |
NM_001195545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007769.hg.1 |
NM_001195545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000578802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007768.hg.1 |
ENST00000582263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007769.hg.1 |
ENST00000377924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007769.hg.1 |
ENST00000377924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007769.hg.1 |
ENST00000377924.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007769.hg.1 |
NM_001195545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007769.hg.1 |
NM_001195545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007769.hg.1 |
NM_001195545.1 |
Refseq |