| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000302273.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
ENST00000403807.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_001303509.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200006768.hg.1 |
NM_007128.3 |
Refseq |