Assay Details
| Gene Symbol: ZNF773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304336.1 | NP_001291265.1 | 3 - 4 | 488 | 103 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000601958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | ENST00000601958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000282292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000593916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000597061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000598770.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000599847.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000601958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | ENST00000601958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304334.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304336.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_001304337.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NM_198542.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011817.hg.1 | NR_130705.1 | Refseq |

ZNF773
Genomic Map