Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304998.1 | NP_001291927.1 | 1 - 2 | 66 | 86 | |
| XM_017022941.1 | XP_016878430.1 | 1 - 2 | 86 | 86 | ||
| GenBank mRNA | DA370427.1 | - | 1 - 2 | 66 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001304998.1, XM_017022941.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000357613.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000561878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000566980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000567796.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000568559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569276.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | ENST00000569540.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304997.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_001304998.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011566.hg.1 | NM_145254.2 | Refseq |

TMEM170A
Genomic Map