Assay Details
| Gene Symbol: CHCHD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317858.1 | NP_001304787.1 | 4 - 5 | 508 | 112 | |
| NM_001317859.1 | NP_001304788.1 | 3 - 4 | 336 | 112 | ||
| GenBank mRNA | BI911444.1 | - | 4 - 5 | 339 | 112 | |
| CN256918.1 | - | 3 - 4 | 166 | 112 | ||
More Information
Set Membership: |
|

CHCHD7
Genomic Map