Assay Details
| Gene Symbol: AP3B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278511.1 | NP_001265440.1 | 22 - 23 | 2968 | 128 | |
| NM_001278512.1 | NP_001265441.1 | 24 - 25 | 3121 | 128 | ||
| NM_004644.4 | NP_004635.2 | 23 - 24 | 3064 | 128 | ||
| XM_017022640.1 | XP_016878129.1 | 22 - 23 | 3196 | 128 | ||
| GenBank mRNA | AF022152.1 | - | 23 - 24 | 2893 | 128 | |
| AK097827.1 | - | 15 - 16 | 1961 | 128 | ||
| BC093739.1 | - | 23 - 24 | 2935 | 7939584 | 128 | |
| BC143342.1 | - | 22 - 23 | 2839 | 9051847 | 128 | |
| BC143346.1 | - | 24 - 25 | 2992 | 9051851 | 128 | |
| CR749860.1 | - | 22 - 23 | 3063 | 128 | ||
| DQ092369.1 | - | 1 - 2 | 509 | 128 | ||
| U37673.1 | - | 23 - 24 | 2920 | 128 | ||
More Information
Set Membership: |
|

AP3B2
Genomic Map