Assay Details
| Gene Symbol: ATP6AP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017010026.1 | XP_016865515.1 | 4 - 5 | 1224 | 120 | |
| XM_017010027.1 | XP_016865516.1 | 4 - 5 | 572 | 120 | ||
| XM_017010028.1 | XP_016865517.1 | 4 - 5 | 857 | 120 | ||
| XM_017010031.1 | XP_016865520.1 | 4 - 5 | 1212 | 120 | ||
| XM_017010032.1 | XP_016865521.1 | 5 - 6 | 1382 | 120 | ||
| XM_017010033.1 | XP_016865522.1 | 4 - 5 | 845 | 120 | ||
| XM_017010041.1 | XP_016865530.1 | 4 - 5 | 1211 | 120 | ||
| XM_017010042.1 | XP_016865531.1 | 4 - 5 | 1211 | 120 | ||
| XM_017010043.1 | XP_016865532.1 | 4 - 5 | 1211 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010026.1, XM_017010027.1, XM_017010028.1, XM_017010031.1, XM_017010032.1, XM_017010033.1, XM_017010041.1, XM_017010042.1, XM_017010043.1 transcripts.
Set Membership: |
|

ATP6AP1L
Genomic Map