Assay Details
| Gene Symbol: ATP6AP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011543719.2 | XP_011542021.1 | 4 - 5 | 1176 | 91 | |
| XM_011543722.2 | XP_011542024.1 | 4 - 5 | 1193 | 91 | ||
| XM_011543724.2 | XP_011542026.1 | 4 - 5 | 1183 | 91 | ||
| XM_017010025.1 | XP_016865514.1 | 4 - 5 | 822 | 91 | ||
| XM_017010026.1 | XP_016865515.1 | 5 - 6 | 1279 | 91 | ||
| XM_017010027.1 | XP_016865516.1 | 5 - 6 | 627 | 91 | ||
| XM_017010028.1 | XP_016865517.1 | 5 - 6 | 912 | 91 | ||
| XM_017010029.1 | XP_016865518.1 | 4 - 5 | 1228 | 91 | ||
| XM_017010030.1 | XP_016865519.1 | 4 - 5 | 861 | 91 | ||
| XM_017010031.1 | XP_016865520.1 | 5 - 6 | 1267 | 91 | ||
| XM_017010032.1 | XP_016865521.1 | 6 - 7 | 1437 | 91 | ||
| XM_017010033.1 | XP_016865522.1 | 5 - 6 | 900 | 91 | ||
| XM_017010034.1 | XP_016865523.1 | 4 - 5 | 1225 | 91 | ||
| XM_017010038.1 | XP_016865527.1 | 4 - 5 | 1181 | 91 | ||
| XM_017010041.1 | XP_016865530.1 | 5 - 6 | 1266 | 91 | ||
| XM_017010042.1 | XP_016865531.1 | 5 - 6 | 1266 | 91 | ||
| XM_017010043.1 | XP_016865532.1 | 5 - 6 | 1266 | 91 | ||
| XM_017010044.1 | XP_016865533.1 | 4 - 5 | 1214 | 91 | ||
| XM_017010046.1 | XP_016865535.1 | 4 - 5 | 847 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017010041.1, XM_017010042.1, XM_017010043.1, XM_017010044.1, XM_017010046.1 transcripts.
Set Membership: |
|

ATP6AP1L
Genomic Map