Assay Details
| Gene Symbol: ATAD3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170535.1 | NP_001164006.1 | 16 | 2317 | 133 | |
| NM_001170536.1 | NP_001164007.1 | 16 | 2147 | 133 | ||
| NM_018188.3 | NP_060658.3 | 16 | 2461 | 133 | ||
| GenBank mRNA | AK001571.1 | - | 16 | 2300 | 133 | |
| AK056099.1 | - | 16 | 2147 | 133 | ||
| AK091144.1 | - | 16 | 2429 | 133 | ||
| AK092833.1 | - | 1 | 2433 | 133 | ||
| BC007803.2 | - | 16 | 2278 | 4136170 | 133 | |
| BC011814.2 | - | 16 | 2283 | 3162989 | 133 | |
| BC014101.2 | - | 16 | 2307 | 3503438 | 133 | |
| BC033109.1 | - | 16 | 2281 | 4877449 | 133 | |
| BC063607.1 | - | 16 | 2275 | 4939339 | 133 | |
More Information
Set Membership: |
|

ATAD3A
Genomic Map