Assay Details
| Gene Symbol: SNORD14E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003125.2 | 1 | 40 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SUMO1 activating enzyme subunit 1 pseudogene | LOC341056 | -38.5kb | Anti-sense | Chr. 11 - 123017566 - 123019611 on NCBI Build GRCh38 | |
| Downstream |
small nucleolar RNA, C/D box 14D |
SNORD14D | 753.0 | Sense | Chr. 11 - 123058909 - 123058995 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORD14E
Genomic Map