Assay Details
| Gene Symbol: MIR4787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_039950.1 | 1 | 40 | 45 | ||
| Gene Symbol: DOCK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005264914.3 | XP_005264971.1 | 1 | 151 | 45 | |
| XM_005264915.3 | XP_005264972.1 | 1 | 151 | 45 | ||
| XM_005264917.3 | XP_005264974.1 | 1 | 151 | 45 | ||
| XM_006713008.3 | XP_006713071.1 | 1 | 151 | 45 | ||
| XM_006713009.3 | XP_006713072.1 | 1 | 151 | 45 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase-activated protein kinase 3 | MAPKAPK3 |
3PK, MAPKAP-K3, MAPKAP3, MAPKAPK-3, MK-3 |
-25.8kb | Sense | Chr. 3 - 50611862 - 50649297 on NCBI Build GRCh38 |
| Downstream | mesencephalic astrocyte derived neurotrophic factor | MANF |
ARMET, ARP |
710.1kb | Sense | Chr. 3 - 51385237 - 51389397 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005264914.3, XM_005264915.3, XM_005264917.3, XM_006713008.3, XM_006713009.3 transcripts.
Set Membership: |
|

MIR4787
Genomic Map