Assay Details
| Gene Symbol: KCND3-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046783.1 | 1 - 2 | 84 | 99 | ||
| GenBank mRNA | BX096614.1 | - | 1 - 2 | 84 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD-box helicase 20 | DDX20 |
DP103, GEMIN3 |
-86.2kb | Anti-sense | Chr. 1 - 111755568 - 111767577 on NCBI Build GRCh38 |
| Downstream | KCND3 antisense RNA 1 | KCND3-AS1 | 52.8kb | Anti-sense | Chr. 1 - 111909657 - 111910930 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

KCND3-IT1
Genomic Map