| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
ENST00000413591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
ENST00000413591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
ENST00000413591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
ENST00000432770.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
ENST00000432770.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
NR_046998.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
NR_046998.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300008508.hg.1 |
NR_046998.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
ENST00000413591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
ENST00000413591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
ENST00000413591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
ENST00000432770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
ENST00000432770.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
NR_046998.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
NR_046998.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300008508.hg.1 |
NR_046998.1 |
Refseq |