Assay Details
| Gene Symbol: LOC105377348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131185.1 | 1 - 2 | 186 | 121 | ||
| GenBank mRNA | HY045516.1 | - | 1 - 2 | 187 | 121 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723704 | LOC102723704 | -29.7kb | Sense | Chr. 4 - 102777037 - 102799575 on NCBI Build GRCh38 | |
| Downstream | CDGSH iron sulfur domain 2 | CISD2 |
ERIS, Miner1, NAF-1, WFS2, ZCD2 |
28.2kb | Sense | Chr. 4 - 102868978 - 102892807 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105377348
Genomic Map