Assay Details
| Gene Symbol: LOC105447648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131191.1 | 1 - 2 | 325 | 87 | ||
| GenBank mRNA | BX458411.2 | - | 1 - 2 | 325 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MIR193B host gene | MIR193BHG | -36.6kb | Anti-sense | Chr. 16 - 14302288 - 14326517 on NCBI Build GRCh38 | |
| Downstream | poly(A)-specific ribonuclease | PARN |
DAN, DKCB6, PFBMFT4 |
65.4kb | Sense | Chr. 16 - 14435700 - 14630277 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105447648
Genomic Map