Assay Details
Gene Symbol: AGAP1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131900.1 | 1 - 2 | 46 | 92 | ||
GenBank mRNA | BX111890.1 | - | 1 - 2 | 46 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1 |
AGAP1 |
AGAP-1, CENTG2, GGAP1, cnt-g2 |
Sense | Chr. 2 - 235494089 - 236131800 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC105373942 | LOC105373942 | 238.7kb | Anti-sense | Chr. 2 - 235746235 - 235791606 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
