Assay Details
| Gene Symbol: LOC102725079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131942.1 | 1 - 2 | 60 | 137 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105373860 | LOC105373860 | -85.9kb | Anti-sense | Chr. 2 - 212706806 - 212709454 on NCBI Build GRCh38 | |
| Downstream | microRNA 4776-1 | MIR4776-1 | mir-4776-1 | 129.5kb | Sense | Chr. 2 - 212926257 - 212926336 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200010687.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175778.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175778.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175778.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175778.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175778.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175778.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175778.hg.1 | TC0200010687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200175782.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175782.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175782.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175782.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175782.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175782.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175782.hg.1 | TC0200010687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200175784.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175784.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175784.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175784.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175784.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175784.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175784.hg.1 | TC0200010687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175775.hg.1 | TC0200010687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200175776.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175776.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175776.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175776.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175776.hg.1 | TC0200010687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000434559.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200010687.hg.1 | NR_131942.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175777.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175777.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175777.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175777.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175777.hg.1 | ENST00000453965.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200175777.hg.1 | NR_131941.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200175777.hg.1 | TC0200010687.hg.1 | Clariom D TC |

LOC102725079
Genomic Map