Assay Details
| Gene Symbol: CRAT40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_131984.1 | 1 - 2 | 175 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | regulator of G-protein signaling 9 | RGS9 |
PERRS, RGS9L |
-229.8kb | Sense | Chr. 17 - 65137338 - 65227703 on NCBI Build GRCh38 |
| Downstream | axin 2 | AXIN2 |
AXIL, ODCRCS |
70.4kb | Anti-sense | Chr. 17 - 65528563 - 65561622 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CRAT40
Genomic Map