Assay Details
| Gene Symbol: SNORD133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132753.1 | 1 | 124 | 110 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 186 member A | FAM186A | -59.9kb | Anti-sense | Chr. 12 - 50326854 - 50396633 on NCBI Build GRCh38 | |
| Downstream | disco interacting protein 2 homolog B | DIP2B | 48.2kb | Sense | Chr. 12 - 50504985 - 50748667 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SNORD133
Genomic Map