Assay Details
| Gene Symbol: LOC101929227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132790.1 | 1 - 2 | 481 | 61 | ||
| GenBank mRNA | CR936773.1 | - | 1 - 2 | 481 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105369532 | LOC105369532 | -48.8kb | Sense | Chr. 11 - 120811345 - 120819115 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929208 | LOC101929208 | 45.8kb | Sense | Chr. 11 - 120940554 - 120958039 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929227
Genomic Map