Assay Details
| Gene Symbol: LOC105378367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132791.1 | 2 - 3 | 400 | 92 | ||
| GenBank mRNA | HY007587.1 | - | 2 - 3 | 401 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 606 | MIR606 |
MIRN606, hsa-mir-606 |
-190.0kb | Sense | Chr. 10 - 75552458 - 75552553 on NCBI Build GRCh38 |
| Downstream | potassium calcium-activated channel subfamily M alpha 1 | KCNMA1 |
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1 |
1.1mb | Anti-sense | Chr. 10 - 76869601 - 77637819 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105378367
Genomic Map