Assay Details
| Gene Symbol: LOC105376599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_132968.1 | 2 - 3 | 270 | 117 | ||
| Gene Symbol: LOC105375847 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX097369.1 | - | 2 - 3 | 270 | 117 | |
More Information
Set Membership: |
|

LOC105376599
Genomic Map