Assay Details
| Gene Symbol: DIRC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_133642.1 | 4 - 5 | 407 | 108 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928278 | LOC101928278 | -52.0kb | Sense | Chr. 2 - 216711707 - 217250714 on NCBI Build GRCh38 | |
| Downstream | tensin 1 | TNS1 |
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS |
463.2kb | Anti-sense | Chr. 2 - 217799789 - 217978853 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

DIRC3-AS1
Genomic Map