Assay Details
| Gene Symbol: ZSCAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_133668.1 | 1 - 2 | 111 | 107 | ||
| XM_011515015.2 | XP_011513317.1 | 4 - 5 | 9141 | 107 | ||
| GenBank mRNA | DB014565.1 | - | 1 - 2 | 75 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger with KRAB and SCAN domains 3 | ZKSCAN3 |
ZF47, ZFP306, ZNF306, ZNF309, ZSCAN13, ZSCAN35, Zfp47, dJ874C20.1, dJ874C20.1., zfp-47 |
-13.0kb | Anti-sense | Chr. 6 - 28349913 - 28369177 on NCBI Build GRCh38 |
| Downstream | zinc finger and SCAN domain containing 23 | ZSCAN23 |
ZNF390, ZNF453, dJ29K1.3, dJ29K1.3.1 |
42.1kb | Sense | Chr. 6 - 28425516 - 28443554 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ZSCAN12
Genomic Map