Assay Details
| Gene Symbol: LOC105379143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134249.1 | 8 - 9 | 2209 | 91 | ||
| GenBank mRNA | BC036311.1 | - | 8 - 9 | 2213 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DTW domain containing 2 | DTWD2 | -6.4kb | Sense | Chr. 5 - 118836074 - 118989938 on NCBI Build GRCh38 | |
| Downstream | Dmx like 1 | DMXL1 | 69.4kb | Anti-sense | Chr. 5 - 119071002 - 119250170 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105379143
Genomic Map