Assay Details
| Gene Symbol: BCL2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134257.1 | 1 - 2 | 679 | 83 | ||
| GenBank mRNA | BX647525.1 | - | 3 - 4 | 557 | 83 | |
| CD359906.1 | - | 1 - 2 | 295 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | apoptotic BCL2L1-antisense long non-coding RNA | ABALON | INXS | Anti-sense | Chr. 20 - 31721507 - 31723409 on NCBI Build GRCh38 | |
| Downstream |
TPX2, microtubule nucleation factor |
TPX2 |
C20orf1, C20orf2, DIL-2, DIL2, FLS353, GD:C20orf1, HCA519, HCTP4, REPP86, p100 |
16.8kb | Anti-sense | Chr. 20 - 31739101 - 31801802 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

BCL2L1
Genomic Map