Assay Details
Gene Symbol: LOC105377763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134260.1 | 2 - 3 | 508 | 94 | ||
GenBank mRNA | HY106435.1 | - | 2 - 3 | 207 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 5 open reading frame 60 | C5orf60 | -13.1kb | Anti-sense | Chr. 5 - 179641542 - 179645167 on NCBI Build GRCh38 | |
Downstream | chibby family member 3 | CBY3 | 3.7kb | Anti-sense | Chr. 5 - 179677969 - 179681830 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
