Assay Details
| Gene Symbol: LOC101927357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134281.1 | 1 - 2 | 2155 | 82 | ||
| GenBank mRNA | BX648985.1 | - | 1 - 2 | 2151 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC153163 | MGC32805 | -102.4kb | Sense | Chr. 5 - 122436497 - 122479087 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927379 | LOC101927379 | 44.4kb | Sense | Chr. 5 - 122628952 - 122730560 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927357
Genomic Map