Assay Details
| Gene Symbol: LOC101927141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134295.1 | 1 - 2 | 274 | 115 | ||
| GenBank mRNA | BX105728.1 | - | 1 - 2 | 274 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorting nexin 16 | SNX16 | -1.1mb | Anti-sense | Chr. 8 - 81799583 - 81842326 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1419 | LINC01419 |
LVCAT7, TCONS_00014497 |
490.7kb | Sense | Chr. 8 - 83403758 - 83408900 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927141
Genomic Map