| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006444.hg.1 |
ENST00000497017.1 |
Ensembl |