| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000462565.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000480919.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000487884.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009958.hg.1 |
ENST00000497320.1 |
Ensembl |