Assay Details
Gene Symbol: LOC102724264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_134483.1 | 1 - 2 | 318 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ZNF32 antisense RNA 3 | ZNF32-AS3 |
C10orf44, ZNF32-OT1, bA402L1.3 |
-102.9kb | Anti-sense | Chr. 10 - 43628817 - 43674699 on NCBI Build GRCh38 |
Downstream | heterogeneous nuclear ribonucleoprotein A3 pseudogene 1 | HNRNPA3P1 |
D10S102, FBRNP, HNRPA3, HNRPA3P1 |
8.5kb | Sense | Chr. 10 - 43787412 - 43790417 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
