Assay Details
| Gene Symbol: LOC105378292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134499.1 | 1 - 2 | 47 | 60 | ||
| GenBank mRNA | BX101243.1 | - | 1 - 2 | 48 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC107001062 | LOC107001062 | Anti-sense | Chr. 10 - 47407165 - 47414574 on NCBI Build GRCh38 | ||
| Downstream | annexin A8 | ANXA8 |
ANX8, CH17-360D5.2 |
53.5kb | Sense | Chr. 10 - 47467993 - 47991879 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105378292
Genomic Map