Assay Details
| Gene Symbol: LOC105379477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134504.1 | 2 - 3 | 326 | 108 | ||
| GenBank mRNA | BC039483.1 | - | 2 - 3 | 326 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
FSHD region gene 2 family member E, pseudogene |
FRG2EP | -38.1kb | Anti-sense | Chr. 20 - 29414075 - 29416961 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1598 | LINC01598 | 90.5kb | Sense | Chr. 20 - 29548719 - 29560475 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105379477
Genomic Map